missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PHLPP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 213.00 - € 508.00
Specifications
| Antigen | PHLPP2 |
|---|---|
| Dilution | Western Blot, ELISA, Immunohistochemistry-Paraffin : 10-15 μg/mL |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066818
|
Novus Biologicals™
NBP3-30962-100ug |
100 μg |
€ 508.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067107
|
Novus Biologicals™
NBP3-30962-25ug |
25 μg |
€ 213.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PHLPP2 Polyclonal antibody specifically detects PHLPP2 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)Specifications
| PHLPP2 | |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Tumor Suppressors | |
| Phosphate buffered saline | |
| 23035 | |
| IgG | |
| Antigen Affinity-purified |
| Western Blot, ELISA, Immunohistochemistry-Paraffin : 10-15 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.1.3.16, KIAA0931PH domain leucine-rich repeat-containing protein phosphatase 2, PH domain and leucine rich repeat protein phosphatase 2, PH domain and leucine rich repeat protein phosphatase-like, PH domain leucine-rich repeat-containing protein phosphatase-like, PHLPPLPHLPP-like | |
| Synthetic peptide derived from the human PHLPP2 protein | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title