missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHLPP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 450.00 - € 708.00
Specifications
| Antigen | PHLPP2 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647484
|
Novus Biologicals
NBP3-21264-25ul |
25 μg |
€ 450.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679744
|
Novus Biologicals
NBP3-21264-100ul |
100 μg |
€ 708.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PHLPP2 Polyclonal antibody specifically detects PHLPP2 in Human samples. It is validated for ImmunofluorescenceSpecifications
| PHLPP2 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Tumor Suppressors | |
| PBS, pH 7.2, 40% glycerol | |
| 23035 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.1.3.16, KIAA0931PH domain leucine-rich repeat-containing protein phosphatase 2, PH domain and leucine rich repeat protein phosphatase 2, PH domain and leucine rich repeat protein phosphatase-like, PH domain leucine-rich repeat-containing protein phosphatase-like, PHLPPLPHLPP-like | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: DLHLVLCTVETPASEICAGEGRESLYLQLHGDLVRRLEPTERPLQIVYDYLSRLGFDDPVRIQEEATNPDLGCMIRFYG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title