Learn More
Phospho-c-Met (Tyr1234/1235) Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30737-1-AP-20UL
 View more versions of this product
Product Code. 18039675
  • € 135.00 / 20µL
Description
The Phospho-c-Met (Tyr1234/1235) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-c-Met (Tyr1234/1235). This antibody recognizes human antigen. The Phospho-c-Met (Tyr1234/1235) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
Phospho-c-Met (Tyr1234/1235)
550 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
AUTS9, c Met, HGF receptor, HGF/SF receptor, HGFR, MET, Proto oncogene c Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine protein kinase Met
20 μL
4233
Store at -20°C. Stable for one year after shipment.