Learn More
Phospho-c-Met (Tyr1234/1235) Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30737-1-AP-20UL
Description
The Phospho-c-Met (Tyr1234/1235) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-c-Met (Tyr1234/1235). This antibody recognizes human antigen. The Phospho-c-Met (Tyr1234/1235) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Phospho-c-Met (Tyr1234/1235) | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| AUTS9, c Met, HGF receptor, HGF/SF receptor, HGFR, MET, Proto oncogene c Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine protein kinase Met | |
| 20 μL | |
| 4233 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |