Learn More
Phospho-c-Met (Tyr1234/1235) Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 135.00 - € 360.00
Specifications
| Antigen | Phospho-c-Met (Tyr1234/1235) |
|---|---|
| Concentration | 550 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18039675
|
Proteintech
30737-1-AP-20UL |
20 μL |
€ 135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18055165
|
Proteintech
30737-1-AP-100UL |
100 μL |
€ 360.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-c-Met (Tyr1234/1235) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-c-Met (Tyr1234/1235). This antibody recognizes human antigen. The Phospho-c-Met (Tyr1234/1235) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Phospho-c-Met (Tyr1234/1235) | |
| Western Blot, ELISA | |
| Rabbit | |
| 4233 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 550 μg/mL | |
| Liquid | |
| Human | |
| AUTS9, c Met, HGF receptor, HGF/SF receptor, HGFR, MET, Proto oncogene c Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine protein kinase Met | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.