missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Caspase 9 (Thr125) Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68136-1-IG-20UL
This item is not returnable.
View return policy
Description
The Phospho-Caspase 9 (Thr125) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-Caspase 9 (Thr125). This antibody recognizes human, rat antigen. The Phospho-Caspase 9 (Thr125) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| Caspase 9 (Thr125) | |
| Monoclonal | |
| 1000 μg/mL | |
| APAF-3; APAF3; CASPASE-9c; ICE-LAP6; MCH6 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| 36 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 1B5E11 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 842 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction