missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Caspase 9 (Thr125) Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 135.00 - € 360.00
Specifications
| Antigen | Caspase 9 (Thr125) |
|---|---|
| Clone | 1B5E11 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233228
|
Proteintech
68136-1-IG-100UL |
100 μL |
€ 360.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234023
|
Proteintech
68136-1-IG-20UL |
20 μL |
€ 135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-Caspase 9 (Thr125) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-Caspase 9 (Thr125). This antibody recognizes human, rat antigen. The Phospho-Caspase 9 (Thr125) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| Caspase 9 (Thr125) | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 842 | |
| Peptide | |
| Primary | |
| Store at -20°C. |
| 1B5E11 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| APAF-3; APAF3; CASPASE-9c; ICE-LAP6; MCH6 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title