Learn More
Phospho-MCM2 (Ser139) Recombinant Rabbit Monoclonal Antibody (MCM2S139-B12), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Thermo Scientific MA537425
 View more versions of this product
Product Code. 17117018
  • € 347.00 / 20µL
Description
MCM2 (Mini chromosome maintenance protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. MCM2 has been mapped to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-MCM2 (Ser139)
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide
P49736, P97310
MCM2
A synthetic phospho-peptide corresponding to residues surrounding Ser139 of human phospho MCM2.
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG κ
Flow Cytometry, Western Blot
MCM2S139-B12
Unconjugated
MCM2
AA959861; AW476101; BM28; CCNL1; cdc19; CDCL1; cell devision cycle-like 1; cyclin-like 1; D3S3194; DFNA70; DNA replication licensing factor MCM2; Kiaa0030; MCM DNA helicase complex subunit MCM2; Mcm2; Mcmd2; MGC10606; mini chromosome maintenance deficient 2; minichromosome maintenance complex component 2; minichromosome maintenance deficient 2 (mitotin); minichromosome maintenance deficient 2 mitotin; minichromosome maintenance deficient 2 mitotin (S. cerevisiae); Minichromosome maintenance protein 2; Minichromosome maintenance protein 2 homolog; MITOTIN; mKIAA0030; Nuclear protein BM28; YBL023C; YBL0438
Rabbit
Protein A/G
RUO
17216, 312538, 4171
-20°C