missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MCM2 (Ser139) Recombinant Rabbit Monoclonal Antibody (MCM2S139-B12), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 331.00 - € 662.00
Specifications
| Antigen | Phospho-MCM2 (Ser139) |
|---|---|
| Clone | MCM2S139-B12 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17117018
|
Thermo Scientific
MA537425 |
20 μL |
€ 331.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17136768
![]() |
Thermo Scientific
MA537177 |
200 μL |
€ 662.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCM2 (Mini chromosome maintenance protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. MCM2 has been mapped to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).Specifications
| Phospho-MCM2 (Ser139) | |
| 0.5 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide | |
| AA959861; AW476101; BM28; CCNL1; cdc19; CDCL1; cell devision cycle-like 1; cyclin-like 1; D3S3194; DFNA70; DNA replication licensing factor MCM2; Kiaa0030; MCM DNA helicase complex subunit MCM2; Mcm2; Mcmd2; MGC10606; mini chromosome maintenance deficient 2; minichromosome maintenance complex component 2; minichromosome maintenance deficient 2 (mitotin); minichromosome maintenance deficient 2 mitotin; minichromosome maintenance deficient 2 mitotin (S. cerevisiae); Minichromosome maintenance protein 2; Minichromosome maintenance protein 2 homolog; MITOTIN; mKIAA0030; Nuclear protein BM28; YBL023C; YBL0438 | |
| MCM2 | |
| IgG κ | |
| Protein A/G | |
| Antibody |
| MCM2S139-B12 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P49736, P97310 | |
| 17216, 312538, 4171 | |
| A synthetic phospho-peptide corresponding to residues surrounding Ser139 of human phospho MCM2. | |
| Primary | |
| -20°C | |
| MCM2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title