missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-S6 Ribosomal protein (Ser236) Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80206-1-RR-20UL
This item is not returnable.
View return policy
Description
The Phospho-S6 Ribosomal protein (Ser236) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-S6 Ribosomal protein (Ser236). This antibody recognizes human, mouse antigen. The Phospho-S6 Ribosomal protein (Ser236) antibody has been validated for the following applications: WB, FC (Intra), ELISA analysis.Specifications
| S6 Ribosomal protein (Ser236) | |
| Recombinant Monoclonal | |
| 250 μg/mL | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 6194 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
| Western Blot, Flow Cytometry (Intracellular Staining), ELISA | |
| 7K17 | |
| Unconjugated | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 32 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction