missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-S6 Ribosomal protein (Ser236) Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
€ 149.00 - € 439.00
Specifications
| Antigen | S6 Ribosomal protein (Ser236) |
|---|---|
| Clone | 7K17 |
| Concentration | 250 μg/mL |
| Applications | Western Blot, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237234
|
Proteintech
80206-1-RR-20UL |
20 μL |
€ 149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234469
|
Proteintech
80206-1-RR-100UL |
100 μL |
€ 439.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-S6 Ribosomal protein (Ser236) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-S6 Ribosomal protein (Ser236). This antibody recognizes human, mouse antigen. The Phospho-S6 Ribosomal protein (Ser236) antibody has been validated for the following applications: WB, FC (Intra), ELISA analysis.Specifications
| S6 Ribosomal protein (Ser236) | |
| 250 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 6194 | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
| 7K17 | |
| Western Blot, Flow Cytometry (Intracellular Staining), ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title