missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PKC beta 1 Antibody (SR3633), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27793-50ul
This item is not returnable.
View return policy
Description
PKC beta 1 Monoclonal antibody specifically detects PKC beta 1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| PKC beta 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5579 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR3633 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| PKCB;PKCbeta;PKC-beta;PKCI(2);PRKCB;PRKCB1;PRKCB2;Protein kinase C beta type | |
| A synthesized peptide derived from human PKC beta 1 (Uniprot #: P05771) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu