missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC zeta, p Thr560 Antibody (1B10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26399-50ul
This item is not returnable.
View return policy
Description
PKC zeta Monoclonal antibody specifically detects PKC zeta in Human samples. It is validated for Western Blot, ELISA
Specifications
| PKC zeta | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 1B10 | |
| Western Blot 1:500-1:5000, ELISA | |
| EC 2.7.11, EC 2.7.11.13, nPKC-zeta, PKC2PKC-ZETA, protein kinase C zeta type, protein kinase C, zeta | |
| A synthesized peptide derived from Human Phospho-PKC zeta [p Thr560] [UniProt Q05513] | |
| 50 μL | |
| Apoptosis, Cancer, Diabetes Research, GPCR, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators | |
| 5590 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction