missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC zeta, p Thr560 Antibody (1B10), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | PKC zeta |
|---|---|
| Clone | 1B10 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30036906
|
Novus Biologicals
NBP3-26399-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30039846
|
Novus Biologicals
NBP3-26399-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKC zeta Monoclonal antibody specifically detects PKC zeta in Human samples. It is validated for Western Blot, ELISASpecifications
| PKC zeta | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11, EC 2.7.11.13, nPKC-zeta, PKC2PKC-ZETA, protein kinase C zeta type, protein kinase C, zeta | |
| A synthesized peptide derived from Human Phospho-PKC zeta [p Thr560] [UniProt Q05513] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 1B10 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Diabetes Research, GPCR, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 5590 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title