Learn More
Novus Biologicals™ PLA2G1B Antibody (Diaccurate patent anti-sPLA2-GIB) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28606-50ug
 View more versions of this product
Product Code. 30046314
  • € 213.00 / 50µg
Description
PLA2G1B Monoclonal antibody specifically detects PLA2G1B in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PLA2G1B
Monoclonal
Unconjugated
P04054
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Diaccurate patent anti-sPLA2-GIB
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Group IB phospholipase A2, Phosphatidylcholine 2-acylhydrolase 1B, phospholipase A2, phospholipase A2, group IB (pancreas), PLA2AMGC119834, PLA2EC 3.1.1.4, PPLA2MGC119835
PLA2G1B
50 μg
Lipid and Metabolism
5319