missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PLA2G1B Antibody (Diaccurate patent anti-sPLA2-GIB) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
€ 213.00 - € 920.00
Specifications
| Antigen | PLA2G1B |
|---|---|
| Clone | Diaccurate patent anti-sPLA2-GIB |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30044592
|
Novus Biologicals™
NBP3-28606-100ug |
100 μg |
€ 338.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047995
|
Novus Biologicals™
NBP3-28606-1mg |
1 mg |
€ 920.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046314
|
Novus Biologicals™
NBP3-28606-50ug |
50 μg |
€ 213.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLA2G1B Monoclonal antibody specifically detects PLA2G1B in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| PLA2G1B | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Lipid and Metabolism | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Group IB phospholipase A2, Phosphatidylcholine 2-acylhydrolase 1B, phospholipase A2, phospholipase A2, group IB (pancreas), PLA2AMGC119834, PLA2EC 3.1.1.4, PPLA2MGC119835 | |
| PLA2G1B | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| Diaccurate patent anti-sPLA2-GIB | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P04054 | |
| 5319 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title