Learn More
PLAGL2 Mouse, anti-Human, Clone: 3C2E5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68606-1-IG-150UL
 View more versions of this product
Product Code. 18088755
  • € 410.00 / 150µL
Description
The PLAGL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PLAGL2. This antibody recognizes human, rabbit antigen. The PLAGL2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PLAGL2
3C2E5
FLJ23283, KIAA0198, PLAGL2, Zinc finger protein PLAGL2
150 μL
5326
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Rabbit
150 kDa