missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAGL2 Mouse, anti-Human, Clone: 3C2E5, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | PLAGL2 |
|---|---|
| Clone | 3C2E5 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18000726
|
Proteintech
68606-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18088755
|
Proteintech
68606-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PLAGL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PLAGL2. This antibody recognizes human, rabbit antigen. The PLAGL2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PLAGL2 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Rabbit | |
| FLJ23283, KIAA0198, PLAGL2, Zinc finger protein PLAGL2 | |
| Primary |
| 3C2E5 | |
| Western Blot, ELISA | |
| Mouse | |
| 5326 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title