Learn More
Poulten & Graf GmbH Plain Pipette for Polystop™ Dropping Bottle
Poulten & Graf GmbH Plain Pipette for Polystop™ Dropping Bottle is for use with a 3 piece stopper.
€ 63.19 - € 70.78
Specifications
Sterility | Non-sterile |
---|---|
Quantity | 10 |
Type | Plain pipette for 3-piece stopper for dropping bottle |
For Use With (Equipment) | Dropping Bottle |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15972290
|
Poulten & Graf GmbH
L188/GP/A |
30 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15982290
|
Poulten & Graf GmbH
L188/GP/B |
50 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15992290
|
Poulten & Graf GmbH
L188/GP/C |
100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- For 3 piece stopper
- For dropping bottles
- Plain
Specifications
Non-sterile | |
10 | |
Plain pipette for 3-piece stopper for dropping bottle | |
Dropping Bottle |