missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasmolipin Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94002-0.02ml
This item is not returnable.
View return policy
Description
Plasmolipin Polyclonal antibody specifically detects Plasmolipin in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Plasmolipin | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Plasma membrane proteolipid, plasmolipin, PMLPTM4SF11plasma membrane proteolipid (plasmolipin), transmembrane 4 superfamily member 11 (plasmolipin) | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PLLP (NP_057077.1). MAEFPSKVSTRTSSPAQGAEASVSALRPDLGFVRSRLGALMLLQLVLGLLVWALIADTPYHLYPAYGWVMFVAVFLWLVTIVLFNLYLFQLHMKLYMVPW | |
| 0.02 mL | |
| Neuroscience | |
| 51090 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction