Learn More
Platinum wire, 0.762mm (0.03in) dia, hard, 99.95% (metals basis)
Product Code. 11337787
Change view
Click to view available options
Quantity:
10 cm
30 cm
50 cm
Unit Size:
10cm
30cm
50cm
Product Code. Quantity unitSize
11337787 10 cm 10cm
11327787 30 cm 30cm
11317787 50 cm 50cm
3 options
Product Code. 11337787
Brand: Thermo Scientific Alfa Aesar 014195.BS
Request Bulk or Custom Format
CAS: 7440-06-4 | Pt | 195.08 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
Specifications
Quantity 10 cm
Packaging Shrink-wrapped
Percent Purity 99.95%
Chemical Name or Material Platinum wire