Learn More
PLAU Monoclonal Antibody (OTI1D4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805314S
This item is not returnable.
View return policy
Description
PLAU Monoclonal antibody specifically detects PLAU in Human samples. It is validated for Western BlotSpecifications
| PLAU | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14994 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9970 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1D4 | |
| Unconjugated | |
| NR1I3 | |
| antagonist of uPA; ATF; BDPLT5; CD87; HGNC:9052; plasminogen activator; plasminogen activator, urinary; plasminogen activator, urokinase; Plau; QPD; uPA; u-PA; UPAM; UPAR; U-plasminogen activator; Urinary plasminogen activator, urokinase; URK; URKR; Urokinase-type plasminogen activator; Urokinase-type plasminogen activator chain B; Urokinase-type plasminogen activator long chain A; Urokinase-type plasminogen activator short chain A | |
| Human recombinant protein fragment corresponding to amino acids 1-236 of human NR1I3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |