missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHA4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57684
This item is not returnable.
View return policy
Description
PLEKHA4 Polyclonal specifically detects PLEKHA4 in Human samples. It is validated for Western Blot.
Specifications
| PLEKHA4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| PEPP-1, PEPP1phosphoinositol 3-phosphate binding protein-1, PH domain-containing family A member 4, phosphoinositol 3-phosphate-binding PH domain protein 1, Phosphoinositol 3-phosphate-binding protein 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 4, pleckstrin homology domain-containing family A member 4, pleckstrin homology domain-containing, family A (phosphoinositide bindingspecific) member 4 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57664 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9H4M7 | |
| PLEKHA4 | |
| Synthetic peptides corresponding to PLEKHA4(pleckstrin homology domain containing, family A member 4) The peptide sequence was selected from the middle region of PLEKHA4. Peptide sequence TEPDSPSPVLQGEESSERESLPESLELSSPRSPETDWGRPPGGDKDLASP. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 92%; Canine: 92%; Pig: 92%; Rabbit: 92%; Equine: 85%; Mouse: 85%; Guinea pig: 84%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction