missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHA4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 466.00
Specifications
| Antigen | PLEKHA4 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
PLEKHA4 Polyclonal specifically detects PLEKHA4 in Human samples. It is validated for Western Blot.Specifications
| PLEKHA4 | |
| Polyclonal | |
| Rabbit | |
| Q9H4M7 | |
| 57664 | |
| Synthetic peptides corresponding to PLEKHA4(pleckstrin homology domain containing, family A member 4) The peptide sequence was selected from the middle region of PLEKHA4. Peptide sequence TEPDSPSPVLQGEESSERESLPESLELSSPRSPETDWGRPPGGDKDLASP. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PEPP-1, PEPP1phosphoinositol 3-phosphate binding protein-1, PH domain-containing family A member 4, phosphoinositol 3-phosphate-binding PH domain protein 1, Phosphoinositol 3-phosphate-binding protein 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 4, pleckstrin homology domain-containing family A member 4, pleckstrin homology domain-containing, family A (phosphoinositide bindingspecific) member 4 | |
| PLEKHA4 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title