missing translation for 'onlineSavingsMsg'
Learn More

PeproTech PlGF-1 Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

€ 219.00 - € 1834.00

Specifications

Antigen PlGF-1
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17825093
GREENER_CHOICE
View Documents
PeproTech
500-P226-50UG
50 μg
€ 219.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17835093
GREENER_CHOICE
View Documents
PeproTech
500-P226-100UG
100 μg
€ 275.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17825163
GREENER_CHOICE
View Documents
PeproTech
500-P226-1MG
1 mg
€ 1834.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AA Sequence of recombinant protein: MLPAVPPQQW ALSAGNGSSE VEVVPFQEVW GRSYCRALER LVDVVSEYPS EVEHMFSPSC VSLLRCTGCC GDENLHCVPV ETANVTMQLL KIRSGDRPSY VELTFSQHVR CECRPLREKM KPERCGDAVP RR. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human PlGF-1. Anti-Human PlGF-1-specific antibody was purified by affinity chromatography employing an immobilized Human PlGF-1 matrix. Sandwich ELISA: To detect Human PlGF-1 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human PlGF-1 (500-P226Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human PlGF-1. Western Blot: To detect Human PlGF-1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human PlGF-1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

PlGF-1 is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-1 is expressed in placental tissues, the colon, and mammary carcinomas. It signals through the VEGFR-1/FLT1 receptor, and stimulates endothelial cell proliferation and migration.
Specifications

Specifications

PlGF-1
ELISA, Western Blot
Unconjugated
Rabbit
Human
P49763
PGF
Primary
-20°C
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
5228
E.coli-derived Recombinant Human PlGF-1.
Antigen affinity chromatography
PGF
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.