missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLGF Antibody (1B6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26432-100ul
This item is not returnable.
View return policy
Description
PLGF Monoclonal antibody specifically detects PLGF in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| PLGF | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 1B6 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| D12S1900, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760 | |
| A synthesized peptide derived from Human PLGF [UniProt P49763] | |
| 100 μL | |
| Angiogenesis | |
| 5228 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction