missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLGF Antibody (1B6), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | PLGF |
|---|---|
| Clone | 1B6 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30024647
|
Novus Biologicals
NBP3-26432-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30033962
|
Novus Biologicals
NBP3-26432-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLGF Monoclonal antibody specifically detects PLGF in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| PLGF | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| D12S1900, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760 | |
| A synthesized peptide derived from Human PLGF [UniProt P49763] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 1B6 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 5228 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title