missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pma1p Monoclonal Antibody (40B7)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547421
This item is not returnable.
View return policy
Description
Pma1p Monoclonal Antibody for Western Blot, ICC/IF
Pma1p is an abundant multidomain protein of yeast which is localized in the plasma membrane. It functions as a major regulator of cytoplasmic pH by pumping protons out of the cell. It is part of the P2 subgroup of cation-transporting ATPases. Since Pma1p is a major plasma membrane protein, antibodies which bind to it are useful markers of yeast plasma membranes.
Specifications
| Pma1p | |
| Monoclonal | |
| Conc. Not Determined | |
| Cell culture media with 5mM sodium azide | |
| P05030 | |
| PMA1 | |
| Yeast nuclear prep. | |
| RUO | |
| 852876 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 40B7 | |
| Unconjugated | |
| PMA1 | |
| PMA1 | |
| Mouse | |
| 500 μL | |
| Primary | |
| Yeast | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction