missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pma1p Monoclonal Antibody (40B7)

Mouse Monoclonal Antibody
€ 303.00 - € 1750.00
Specifications
| Antigen | Pma1p |
|---|---|
| Clone | 40B7 |
| Concentration | Conc. Not Determined |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17895047
|
Invitrogen™
MA547323 |
2.5 mL |
€ 1750.00
2.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17835127
|
Invitrogen™
MA547421 |
500 μL |
€ 504.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17825207
![]() |
Invitrogen™
MA547422 |
250 μL |
€ 303.00
250µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pma1p Monoclonal Antibody for Western Blot, ICC/IF
Pma1p is an abundant multidomain protein of yeast which is localized in the plasma membrane. It functions as a major regulator of cytoplasmic pH by pumping protons out of the cell. It is part of the P2 subgroup of cation-transporting ATPases. Since Pma1p is a major plasma membrane protein, antibodies which bind to it are useful markers of yeast plasma membranes.Specifications
| Pma1p | |
| Conc. Not Determined | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| Cell culture media with 5mM sodium azide | |
| P05030 | |
| 852876 | |
| Yeast nuclear prep. | |
| Antibody |
| 40B7 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG | |
| Yeast | |
| PMA1 | |
| PMA1 | |
| PMA1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title