missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PMEL Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

€ 477.00 - € 1204.00

Specifications

Antigen PMEL Biosimilar
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30284598
GREENER_CHOICE
View Documents
Invitrogen™
MA558381
100 μg
€ 477.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30284029
View Documents
Invitrogen™
MA558382
1 mg
€ 1204.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Genentech anti-PMEL17.

The Premelanosome protein (PMEL) gene, also known as PMEL17, plays a critical role in the formation of pigment cells. It is located on chromosome 12 and codes for a protein critical for melanosome biogenesis, the organelles responsible for melanin production. PMEL forms amyloid fibrils, which serve as a scaffold for melanin deposition, thus influencing pigmentation. The structure includes several domains, particularly a repeat (RPT) domain that governs the formation of fibrillar sheets. Mutations in PMEL can lead to pigmentation disorders and have been associated with conditions such as pigmentary glaucoma. Additionally, its expression and functionality are crucial in various species and can influence pigmentation patterns. The gene's evolutionary history highlights unique functional domains that contribute to its role in pigment cell biology.
Specifications

Specifications

PMEL Biosimilar
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.