missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PMP70 Recombinant Rabbit Monoclonal Antibody (23GB5010), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552899
This item is not returnable.
View return policy
Description
Peroxisomes are single membrane organelles which are involved with many important biochemical pathways and are found in almost all eukaryotic cells. Various toxins including, phenols, alcohols, and aldehydes are targeted for modification by peroxisomes. Long chain fatty acid metabolism via beta-oxidation is also mediated by this organelle. Loss of peroxisomal functions may result in diseases such as Zellweger syndrome, hyperpipecolic acidemia, neonatal adrenoleukodystrophy, and infantile Refsum' disease. Peroxisomal membrane protein 70 (PMP70) is an abundant, integral membrane protein of the peroxisome. This protein is induced by treatment with hypolipidemic agents in parallel with peroxisome proliferation and stimulation of the peroxisomal beta-oxidation enzymes.
Specifications
| PMP70 | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P16970, P28288, P55096 | |
| ABCD3 | |
| A synthesized peptide derived from human PMP70 (620-659AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5010 | |
| Unconjugated | |
| ABCD3 | |
| 68 kDa peroxisomal membrane protein; 70 kDa peroxisomal membrane protein; 70-kDa peroxisomal membrane protein; ABC43; ABCD3; AI313901; ATP binding cassette subfamily D member 3; ATP-binding cassette sub-family D member 3; ATP-binding cassette, subfamily D (ALD), member 3; ATP-binding cassette, sub-family D (ALD), member 3; AU018866; AW146054; CBAS5; dJ824O18.1 (ATP-binding cassette, sub-family D (ALD), member 3 (PMP70, PXMP1)); Peroxisomal membrane protein 1; peroxisomal membrane protein 1 (70kD, Zellweger syndrome); peroxisomal membrane protein, 70 kDa; Peroxisomal membrane protein-1 (70kD); PMP68; Pmp70; Pxmp1; ZWS2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19299, 25270, 5825 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction