Learn More
PMS1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94782-0.1ml
This item is not returnable.
View return policy
Description
PMS1 Polyclonal antibody specifically detects PMS1 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| PMS1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp781M0253, FLJ98259, HNPCC3, hPMS1, human homolog of yeast mutL, mismatch repair gene PMSL1, PMS1 postmeiotic segregation increased 1 (S. cerevisiae), PMS1 protein homolog 1, PMSL1DNA mismatch repair protein PMS1, postmeiotic segregation increased (S. cerevisiae) 1, rhabdomyosarcoma antigen MU-RMS-40.10B, rhabdomyosarcoma antigen MU-RMS-40.10E | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 405-619 of human PMS1 (NP_000525.1). DCLNHQISIGDFGYGHCSSEISNIDKNTKNAFQDISMSNVSWENSQTEYSKTCFISSVKHTQSENGNKDHIDESGENEEEAGLENSSEISADEWSRGNILKNSVGENIEPVKILVPEKSLPCKVSNNNYPIPEQMNLNEDSCNKKSNVIDNKSGKVTAYDLLSNRVIKKPMSASALFVQDHRPQFLIENPKTSLEDATLQIEELWKTLSEEEKLK | |
| 0.1 mL | |
| DNA Repair, Mismatch Repair, Ovarian Carcinoma Cell Markers | |
| 5378 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |