missing translation for 'onlineSavingsMsg'
Learn More

PMS1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 206.00 - € 470.00

Specifications

Antigen PMS1
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18663592
View Documents
Novus Biologicals
NBP2-94782-0.02ml
0.02 mL
€ 206.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18683391
View Documents
Novus Biologicals
NBP2-94782-0.1ml
0.1 mL
€ 470.00
0.01mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PMS1 Polyclonal antibody specifically detects PMS1 in Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

PMS1
Western Blot
Unconjugated
Rabbit
DNA Repair, Mismatch Repair, Ovarian Carcinoma Cell Markers
PBS (pH 7.3), 50% glycerol
5378
IgG
Affinity purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Mouse, Rat
DKFZp781M0253, FLJ98259, HNPCC3, hPMS1, human homolog of yeast mutL, mismatch repair gene PMSL1, PMS1 postmeiotic segregation increased 1 (S. cerevisiae), PMS1 protein homolog 1, PMSL1DNA mismatch repair protein PMS1, postmeiotic segregation increased (S. cerevisiae) 1, rhabdomyosarcoma antigen MU-RMS-40.10B, rhabdomyosarcoma antigen MU-RMS-40.10E
Recombinant fusion protein containing a sequence corresponding to amino acids 405-619 of human PMS1 (NP_000525.1). DCLNHQISIGDFGYGHCSSEISNIDKNTKNAFQDISMSNVSWENSQTEYSKTCFISSVKHTQSENGNKDHIDESGENEEEAGLENSSEISADEWSRGNILKNSVGENIEPVKILVPEKSLPCKVSNNNYPIPEQMNLNEDSCNKKSNVIDNKSGKVTAYDLLSNRVIKKPMSASALFVQDHRPQFLIENPKTSLEDATLQIEELWKTLSEEEKLK
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.