missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poliovirus Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-63038
This item is not returnable.
View return policy
Description
Poliovirus Polyclonal specifically detects Poliovirus in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Poliovirus | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) with 0.1% Sodium Azide | |
| Goat | |
| IgG purified | |
| RUO | |
| Specific for poliovirus. The will recognize poliovirus types 1, 2 and 3. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 4.0 mg/mL | |
| ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Enterovirus C, poliomyelitis | |
| Pool of purified poliovirus (types 1-3) | |
| 0.5 mL | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction