missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poliovirus Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
€ 212.00 - € 486.00
Specifications
| Antigen | Poliovirus |
|---|---|
| Concentration | 4.0 mg/mL |
| Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | ELISA, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18087133
|
Novus Biologicals
NB100-63038-0.1ML |
0.1 mL |
€ 212.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18705583
|
Novus Biologicals
NB100-63038 |
0.5 mL |
€ 486.00
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Poliovirus Polyclonal specifically detects Poliovirus in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Poliovirus | |
| ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) with 0.1% Sodium Azide | |
| IgG | |
| IgG purified | |
| Specific for poliovirus. The will recognize poliovirus types 1, 2 and 3. |
| 4.0 mg/mL | |
| ELISA, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Goat | |
| Virus | |
| Pool of purified poliovirus (types 1-3) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title