missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR3D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 624.00
Specifications
| Antigen | POLR3D |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630137
|
Novus Biologicals
NBP2-49530-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647536
|
Novus Biologicals
NBP2-49530 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
POLR3D Polyclonal antibody specifically detects POLR3D in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| POLR3D | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| BN51, BN51 (BHK21) temperature sensitivity complementing, BN51TDNA-directed RNA polymerase III 47 kDa polypeptide, DNA-directed RNA polymerase III subunit D, DNA-directed RNA polymerase III subunit RPC4, polymerase (RNA) III (DNA directed) polypeptide D, 44kDa, Protein BN51, RNA polymerase III 47 kDa subunit, RNA polymerase III subunit C4, RPC4, RPC53 homolog, temperature sensitive complementation, cell cycle specific, tsBN51, TSBN51RPC53 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LPKDVSVAELLRELSLTKEEELLFLQLPDTLPGQPPTQDIKPIKTEVQGEDGQVVLIKQE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| 661 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title