Learn More
Fisherbrand™ Laser Tough-Spots™ Label Sheets
Adhere to most plastics, glass, and metals without cracking, peeling or degrading. Fisherbrand™ Laser Tough-Spots™ Label Sheets are made with flexible polyolefin material and acrylic adhesive.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Resistant to most solvents and caustic agents.
- Label sheets will not jam printers.
- Flexible polyolefin material and acrylic adhesive
- On sheets - 20 sheets per pack
- Rainbow packs contain 4 sheets of each color
- Autoclavable
- Disposable
Specifications
Autoclavable | |
White | |
192 | |
9.5 mm |
Acrylic | |
12 | |
3,840/PK |