missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ Polyethylene Flange Covers, GPN 670 Series

Prevents accidental stripping. Poppelmann™ Polyethylene Flange Covers, GPN 670 Series larger tolerances accomodated by ribs.

Brand:  Poeppelmann™ 67001000000

Additional Details : Weight : 1.23300kg

 View more versions of this product

Product Code. 10247064

  • € 69.00 / Pack of 25
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

  • Polyethylene
  • Yellow
Specifications

Specifications

87 x 109 mm
Flange Covers
25 Pack
BASF-Werknorm 14-150. and screw holes
Closes the opening and covers sealing face and internal threads. Prevents accidental dislodging
Yellow
Polyethylene
24 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers