missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ Polyethylene Flange Covers, GPN 670 Series

Prevents accidental stripping. Poppelmann™ Polyethylene Flange Covers, GPN 670 Series larger tolerances accomodated by ribs.

Brand:  Poeppelmann™ 67000800000

Additional Details : Weight : 1.93500kg

 View more versions of this product

Product Code. 10597364

  • € 124.00 / Pack of 50
Estimated Shipment: 27-06-2024
to see stock.
Add to basket

Description

Description

  • Polyethylene
  • Yellow
Specifications

Specifications

65 x 85 mm
Flange Covers
50 Pack
BASF-Werknorm 14-150. and screw holes
Closes the opening and covers sealing face and internal threads. Prevents accidental dislodging
Yellow
Polyethylene
24 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers