Description
- Polypropylene graduated tubes
- Autoclavable
- Available in a variety of colors for easy identification
- Available with pick-up tab
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 1.5 mL |
| Color | Natural |
| Material | Polypropylene |
| For Use With (Application) | For PCR, Centrifugation |
| No. per Pack | 500/Pk. |
| Graduated | Yes |
| Closure Type | Snap Cap |