Learn More
JG Finneran Associates™ Polypropylene Screw Caps
Brand:  JG Finneran Associates™ 5320-10
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12442980
  • € 67.85 / Pack of 1000
Estimated Shipment: 12-06-2024
to see stock.
Specifications
Linerless
Polypropylene
1000 Pk.
10-425
Black
Molded
Unlined Solid Top Screw Cap
Autosamplers