Learn More
Honeywell™ POLYTRIL™ Air Comfort Gloves
Description
- Polyamide/Cotton/Elasthane knitted glove
- Black foam nitrile coating on palm and fingers
- The ergonomic seamless knit construction conforms better to the natural shape of the hand for greater strength and durability
- The Lycra™ yarn gives a better tactile sensitivity for the assembling of small parts
- The addition of cotton increases the absorption of perspiration and comfort
- Nitrile foam coating and open back provide superior ventilation of the hand
- The liner combining polyamide/cotton/elasthanne avoids skin contact with the coating and helps keeping hands cool and dry
- The nitrile coating provides oil and abrasion resistance and breathability
- The foam version of nitrile helps improving the grip in wet and oily environments
- Black color suitable for dirty applications and environments
Specifications
Specifications
| Certifications/Compliance | EN 420-03 Basic standards, EN 388-16 Mechanical hazards |
| Description | 13 gauge |
| Coating Material | Polyamide, Cotton, Lycra |
| Color | Black |
| Size | 8 |
| Length (Metric) | 24.5 cm |