Learn More
Honeywell™ POLYTRIL™ Air Comfort Gloves
Product Code. 15826725
Change view
Click to view available options
Size:
6
7
8
9
Unit Size:
1 pair
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15826725 8 1 pair
15806725 6 1 pair
15816725 7 1 pair
15836725 9 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15826725 Supplier Honeywell™ Supplier No. 223227308
Only null left
Add to basket
Edge
Add to basket
Honeywell™ POLYTRIL™ Air Comfort Gloves for light handling, assembling in an oily environment, handling in a wet and greasy environment, and assembly work.
  • Polyamide/Cotton/Elasthane knitted glove
  • Black foam nitrile coating on palm and fingers
  • The ergonomic seamless knit construction conforms better to the natural shape of the hand for greater strength and durability
  • The Lycra™ yarn gives a better tactile sensitivity for the assembling of small parts
  • The addition of cotton increases the absorption of perspiration and comfort
  • Nitrile foam coating and open back provide superior ventilation of the hand
  • The liner combining polyamide/cotton/elasthanne avoids skin contact with the coating and helps keeping hands cool and dry
  • The nitrile coating provides oil and abrasion resistance and breathability
  • The foam version of nitrile helps improving the grip in wet and oily environments
  • Black color suitable for dirty applications and environments
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-03 Basic standards, EN 388-16 Mechanical hazards
Description 13 gauge
Coating Material Polyamide, Cotton, Lycra
Color Black
Size 8
Length (Metric) 24.5 cm