Learn More
Honeywell™ POLYTRIL™ Black Gloves
Product Code. p-7159522
Change view
Click to view available options
Size:
6
7
8
9
Unit Size:
1 pair
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15836705 6 1 pair
15846705 7 1 pair
15856705 8 1 pair
15866705 9 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15836705 Supplier Honeywell™ Supplier No. 223223106
Only null left
Add to basket
Add to basket
Honeywell™ POLYTRIL™ Black Gloves for handling of fine objects in a greasy, damp or soiled atmosphere.
  • Black polyamide knitted glove
  • Black oil proof nitrile coating on palm and fingertips
  • The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The ventilated back and PU coating optimise the glove's breathability
  • The knitted and elasticated wrist gives the glove good support
  • The filament yarn reduce linting for the most sensitive applications and easily laundered for extended use
  • The nitrile coating at the palm and fingertips provides outstanding abrasion resistance and impermeability to oils
  • Its dark color makes it more soilproof, thus ensuring a longer life
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-03 Basic standards, EN 388-16 Mechanical hazards
Description 13 gauge
Coating Material Polyamide, Nitrile
Color Black
Size 6
Length (Metric) 23 cm