Learn More
Honeywell™ POLYTRIL™ Mix Gloves
Product Code. 15806715
Change view
Click to view available options
Size:
6
7
8
9
Unit Size:
1 pair
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15806715 9 1 pair
15876705 6 1 pair
15886705 7 1 pair
15896705 8 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15806715 Supplier Honeywell™ Supplier No. 223223309
Only null left
Add to basket
Edge
Add to basket
Honeywell™ POLYTRIL™ Mix Gloves for handling heavy and sharp objects in a greasy, damp and/or dirty environment.
  • Black polyamide/cotton knitted glove
  • Rough black nitrile coating on palm and fingers
  • The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The ventilated back and PU coating optimise the glove's breathability
  • The knitted and elasticated wrist gives the glove good support
  • The cotton polyamide thread offers excellent comfort and provides the glove with effective properties against cuts
  • The nitrile coating at the palm and fingers provides outstanding abrasion resistance and impermeability to oils
  • Its black color makes it more soilproof, thus ensuring a longer life
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE CATEGORY : II Intermediary risks In accordance with standards: EN 420-03 Basic standards EN 388-16 Mechanical hazards
Description 10 gauge
Coating Material Polyamide
Color Black
Size 9
Length (Metric) 25.5 cm