missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hexacyanoferrate(Iii), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
500 g
1 kg
6 x 1 kg
Unit Size:
1kg
250g
500g
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 6 x 1 kg |
| Linear Formula | K3Fe(CN)6 |
| IUPAC Name | tripotassium hexacyanoirontris(ylium) |
| Formula Weight | 329.24 |
| Percent Purity | ≥99% |
| Grade | ACS/puriss. p.a. |
| Chemical Name or Material | Potassium Hexacyanoferrate(Iii), Reag. Ph. Eur. reag. ISO |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction