missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide 50%, For CO2 Determination, Kraft™

Product Code. 10074121
Click to view available options
Quantity:
1 L
Packaging:
Bottle
missing translation for 'unitSize'
1 L
This item is not returnable. View return policy

Product Code. 10074121

missing translation for 'mfr': Kraft 50503000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For determination of carbon dioxide according to Orsat.

  • Molecular Formula: KOH
  • Molar Mass: 56.11g/mol
  • CAS: 1310-58-3

Specifications

Boiling Point 146°C
Melting Point 10°C
CAS 1310-58-3, 7732-18-5
Color Colorless
Physical Form Liquid
Quantity 1 L
Molecular Formula HKO
MDL Number MFCD00003553
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11g/mol
Packaging Bottle
Density 1.510g/cm3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.