Learn More
Potassium Iodide Solution, EU Pharmacopoeia Grade, Fisher Chemical™
BIOPHARMA
Product Code. 12930884 Shop All Fisher Chemical Products
Click to view available options
:
Quantity: 1000mL
Product Code. 12930884
Brand: Fisher Chemical VL 045 1000
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Quantity 1000 mL