Learn More
Potassium Permanganate Solution, 0.02M, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | 0.02mol/L |
Safety and Handling
Product Identifier
- Potassium permanganate 0.2M Volumetric Solution (SOL 119)
- LONG-TERM AQUATIC HAZARD Chronic 2
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to .