missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 49% min

Product Code. 11306528
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11306528

Brand: Thermo Scientific Alfa Aesar 012150.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27988-75-6 | AuCl4K | 377.86 g/mol

Potassium tetrachloroaurate(III) hydrate is used as a raw material for the preparation of gold(III) dithiolate complexes, which finds application in luminescence studies, photosensitizers and photocatalysts. Further, it is used for gold electroplating process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27988-75-6
Molecular Formula AuCl4K
Molecular Weight (g/mol) 377.86
MDL Number MFCD00043088
InChI Key IKHCGJWURGSQRT-UHFFFAOYSA-J
Synonym potassium tetrachloroaurate iii hydrate, gold potassium chloride, aucl4.k.h2o, potassium hydrate tetrachloroaurate, potassium tetrachloroaurate iii hydrate, premion
PubChem CID 91886546
IUPAC Name potassium;tetrachlorogold(1-);hydrate
SMILES [K+].Cl[Au+3](Cl)(Cl)Cl

Specifications

Melting Point 357°C (decomposition)
Color Yellow to Orange
Physical Form Crystalline
Quantity 5 g
Linear Formula KAuCl4·xH2O
UN Number UN3260
Merck Index 14,7681
Solubility Information Soluble in water.
Formula Weight 377.88 (Anhydrous)
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Au 49% min.
Assay (metals basis)
Chemical Name or Material Potassium tetrachloroaurate(III) hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.