Learn More
Honeywell™ POWERCOAT™ 950-30 Neofit Gloves
Click to view available options
Size:
7
8
9
Packungsgröße:
1 pair
Beschreibung
- Neoprene offers high flexibility
- The cotton interlock lining strengthens the glove's mechanical performance while absorbing hand perspiration for added comfort
- The rough finish provides a better grip in a damp environment
- Neoprene provides chemical and mechanical resistance
- It resists acids, bases, solvents as well as industrial adhesives
- Its double coating ensures a good level of chemical protection and superior mechanical protection
- The cuff helps to protect a large part of the forearm
- Supported Neoprene glove, double dipped
- Lined on a 100% cotton interlock cut and sewn liner
- Rough finishing
Spezifikation
Spezifikation
| Certifications/Compliance | EN 420-2003 EEC Basic standard, EN 388-2003 Mechanical risks, EN 374-2003 Chemical risks |
| Color | Black, Blue |
| Material | Neoprene |
| Size | 7 |
| Thickness (Metric) | 1.1 mm |
| Length (Metric) | 30 cm |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur