missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP2C alpha/PPM1A Antibody (7F12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37480-0.025ml
This item is not returnable.
View return policy
Description
PP2C alpha/PPM1A Monoclonal specifically detects PP2C alpha/PPM1A in Human, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| PP2C alpha/PPM1A | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| EC 3.1.3.16, FLJ42306, MGC9201, phosphatase 2C alpha, PP2CA, PP2Calpha, PP2C-ALPHA, PPPM1A, protein phosphatase 1A, protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform, protein phosphatase 2C alpha isoform, Protein phosphatase 2C isoform alpha, Protein phosphatase IA, protein phosphatase, Mg2+/Mn2+ dependent, 1A | |
| Mouse | |
| 42.4 kDa | |
| 0.025 mL | |
| Signal Transduction | |
| 5494 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 7F12 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PPM1A | |
| Purified recombinant fragment of human PPM1A (AA: 202-382) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Primate, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction